missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX1 Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-282JF549
This item is not returnable.
View return policy
Description
SNX1 Polyclonal antibody specifically detects SNX1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Immunohistochemistry, Immunofluorescence | |
| 50mM Sodium Borate | |
| Immunogen affinity purified | |
| 6642 | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| A synthetic peptide, which represented a portion of human Sorting Nexin 1 encoded within exon 15 (LocusLink ID 6642). | |
| Primary | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction