missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20441
This item is not returnable.
View return policy
Description
SNX1 Polyclonal specifically detects SNX1 in Human samples. It is validated for Western Blot.
Specifications
| SNX1 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| HsT17379, MGC8664, SNX1Asorting nexin 1A, sorting nexin 1, sorting nexin-1, Vps5 | |
| Rabbit | |
| 59 kDa | |
| 0.1 mL | |
| Signal Transduction | |
| 6642 | |
| Human | |
| IgG |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q13596 | |
| SNX1 | |
| Recombinant protein encompassing a sequence within the center region of human SNX1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Species reactivity based on sequence identity: Rhesus Monkey (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction