missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNX1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA519239
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This endosomal protein regulates the cell-surface expression of epidermal growth factor receptor. This protein also has a role in sorting protease-activated receptor-1 from early endosomes to lysosomes. This protein may form oligomeric complexes with family members. This gene results in three transcript variants encoding distinct isoforms.
Specifications
| SNX1 | |
| Polyclonal | |
| Unconjugated | |
| SNX1 | |
| HsT17379; MGC8664; Snx1; SNX1A; sorting nexin 1; sorting nexin 1A; sorting nexin-1; VPS5 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 6642 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q13596 | |
| SNX1 | |
| Synthetic peptide sequence (TNSSKPQPTYEELE) corresponding to the internal amino acids of SNX1 (aa 122-136). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction