Learn More
SNX10 Antibody (OTI6B7), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-74262JF646
Product Code. 18168385
  • ONLINE EXCLUSIVE PRICE
    € 785.00
    € 742.35 / 0.10mL
    Save € 42.65
    5% Off
Description
SNX10 Monoclonal specifically detects SNX10 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SNX10
Monoclonal
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
SNX10
Full length recombinant protein of human SNX10 (NP_037454) produced in E.coli.
0.1 mL
Primary
Human, Mouse, Rat
IgG1
Western Blot, Immunohistochemistry
OTI6B7
Western Blot, Immunohistochemistry
MGC33054, sorting nexin 10, sorting nexin-10
Mouse
Affinity Purified
Signal Transduction
29887.0
Store at 4°C in the dark.