missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX10 Monoclonal Antibody (OTI3F5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808860S
This item is not returnable.
View return policy
Description
SNX10 Monoclonal antibody specifically detects SNX10 in Human samples. It is validated for Western BlotSpecifications
| SNX10 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P28906 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 947 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI3F5 | |
| Unconjugated | |
| Cd34 | |
| 2410004M09Rik; OPTB8; Snx10; sorting nexin 10; sorting nexin-10; Sorting nexin-10-like protein | |
| Human recombinant protein fragment corresponding to amino acids 312-385 of human CD34 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction