Learn More
SNX10 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26727-1-AP-20UL
 View more versions of this product
Product Code. 18046895
  • € 145.00 / 20µL
Description
The SNX10 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SNX10. This antibody recognizes human antigen. The SNX10 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
SNX10
1000 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, Immunohistochemistry, ELISA
SNX10, sorting nexin 10
20 μL
29887
Store at -20°C. Stable for one year after shipment.