missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNX11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560827
This item is not returnable.
View return policy
Description
Immunogen sequence: LFLQSQLSVP EIEACVQGRS TMTVSDAILR YAMSNCGWAQ EERQSSSHLA KGDQPKSCCF LPRSGRRSSP Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members. This gene encodes a protein of unknown function. This gene results in two transcript variants differing in the 5' UTR, but encoding the same protein.
Specifications
| SNX11 | |
| Polyclonal | |
| Unconjugated | |
| SNX11 | |
| 4933439F10Rik; A930041K09Rik; AI117694; MGC111019; SNX11; sorting nexin 11; sorting nexin-11 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29916, 303493, 74479 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q91WL6, Q9Y5W9 | |
| SNX11 | |
| Recombinant protein corresponding to Human SNX11. Recombinant protein control fragment (Product #RP-100378). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction