missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX12 Antibody (OTI3A4), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74263CL1
This item is not returnable.
View return policy
Description
SNX12 Monoclonal antibody specifically detects SNX12 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| SNX12 | |
| Monoclonal | |
| CoraFluor 1 | |
| B-cell CLL/lymphoma 7 protein family member C, B-cell CLL/lymphoma 7C | |
| Full length human recombinant protein of human SNX12 (NP_037478) produced in E.coli. | |
| 0.1 mL | |
| Signal Transduction | |
| 29934 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| OTI3A4 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction