missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX12 Monoclonal Antibody (OTI8B3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805077
This item is not returnable.
View return policy
Description
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members. A similar protein in mouse may be involved in regulating the neurite outgrowth. Alternate splicing results in multiple transcript variants.Specifications
| SNX12 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UMY4 | |
| SNX12 | |
| Full length human recombit protein of human SNX12 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI8B3 | |
| Unconjugated | |
| SNX12 | |
| sorting nexin 12 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 29934 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction