missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX12 Monoclonal Antibody (OTI8B3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805077S
This item is not returnable.
View return policy
Description
SNX12 Monoclonal antibody specifically detects SNX12 in Human samples. It is validated for Western BlotSpecifications
| SNX12 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P41250 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2617 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI8B3 | |
| Unconjugated | |
| GARS1 | |
| 2610001F05Rik; AW045757; fc02c04; fc03c06; fj97e07; LOW QUALITY PROTEIN: sorting nexin-12; MGC118982; MGC118983; RGD1565585; SDP8; SDP8 protein; similar to sorting nexin 12; snx12; sorting nexin 12; sorting nexin 12, variation 1; sorting nexin 12, variation 2; sorting nexin-12; wu:fc02c04; wu:fc03c06; wu:fj97e07; zgc:158590; zgc:55728; zgc:77034; zgc:77034 protein | |
| Full length human recombinant protein of human GARS produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction