missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNX13 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121589
This item is not returnable.
View return policy
Description
SNX13 Polyclonal Antibody for Western Blot, IHC (P)
May be involved in several stages of intracellular trafficking. May play a role in endosome homeostasis. Acts as a GAP for Galphas.
Specifications
| SNX13 | |
| Polyclonal | |
| Unconjugated | |
| SNX13 | |
| KIAA0713; mKIAA0713; RGS domain- and PHOX domain-containing protein; RGS-PX1; Snx13; sorting nexin 13; sorting nexin-13 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 23161 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9Y5W8 | |
| SNX13 | |
| Recombinant protein Sorting Nexin 13. The antigen corresponds to amino acid range 97-284 of the target protein. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction