missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX16 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18461UV
This item is not returnable.
View return policy
Description
SNX16 Polyclonal antibody specifically detects SNX16 in Human samples. It is validated for Western Blot
Specifications
| SNX16 | |
| Polyclonal | |
| Western Blot | |
| DKFZp666H147, sorting nexin 16, sorting nexin-16 | |
| Recognizes region between residues Between Between 250 and 300.. | |
| 0.1 mL | |
| Signal Transduction | |
| 64089 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction