missing translation for 'onlineSavingsMsg'
Learn More

SNX17 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 190.00 - € 470.00

Specifications

Antigen SNX17
Dilution Western Blot 1:1000-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18667501
View Documents
Novus Biologicals
NBP2-93381-0.02ml
0.02 mL
€ 190.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18665611
View Documents
Novus Biologicals
NBP2-93381-0.1ml
0.1 mL
€ 470.00
0.01mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SNX17 Polyclonal antibody specifically detects SNX17 in Human samples. It is validated for Western Blot
Specifications

Specifications

SNX17
Western Blot
Unconjugated
Rabbit
Signal Transduction
PBS (pH 7.3), 50% glycerol
9784
IgG
Affinity purified
Western Blot 1:1000-1:2000
Polyclonal
Purified
RUO
Human
KIAA0064sorting nexin-17, sorting nexin 17
Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human SNX17 (NP_055563.1). MHFSIPETESRSGDSGGSAYVAYNIHVNGVLHCRVRYSQLLGLHEQLRKEYGANVLPAFPPKKLFSLTPAEVEQRREQLEKYMQAVRQDPLLGSSETFNSFLRRAQQETQQVPTEEVSLEVLLSNGQKVLVNVLTSDQTEDVLEAVAAKLDLPDDLIGYFSLFLVREKEDGAFSFVRKLQEFELPYVSVTSLRSQEYKIVLRKSYWDSAYDDDVMENRVGLNLLYAQTVSDIERGWILVTKEQHRQLKSLQEKVSKKEFLRLAQTLRHYG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.