missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX18 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93449-0.1ml
This item is not returnable.
View return policy
Description
SNX18 Polyclonal antibody specifically detects SNX18 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| SNX18 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| FLJ11997, FLJ61062, MGC150827, MGC150829, SH3 and PX domain-containing protein 3B, SH3PX2, SH3PXD3BFLJ32560, SNAG1, sorting nexin 18, sorting nexin associated golgi protein 1, sorting nexin-18, Sorting nexin-associated Golgi protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 445-624 of human SNX18 (NP_001096045.1). LNHTANEFARKQVTGFKKEYQKVGQSFRGLSQAFELDQQAFSVGLNQAIAFTGDAYDAIGELFAEQPRQDLDPVMDLLALYQGHLANFPDIIHVQKGALTKVKESRRHVEEGKMEVQKADGIQDRCNTISFATLAEIHHFHQIRVRDFKSQMQHFLQQQIIFFQKVTQKLEEALHKYDSV | |
| 0.1 mL | |
| Signal Transduction | |
| 112574 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction