missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SNX18 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17151824 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17151824 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17151824 Supplier Invitrogen™ Supplier No. PA5113965

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LLQPQQAPPP STFQPPGAGF PYGGGALQPS PQQLYGGYQA SQGSDDDWDD For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 88%; Dog: 80%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Rat: 93%; Yeast: 77%.

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members, but contains a SH3 domain. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX18
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SNX18
Gene Accession No. Q96RF0
Gene Alias SH3 and PX domain-containing protein 3B; SH3PX2; SH3PXD3B; Snag1; Snx18; sorting nexin 18; sorting nexin associated golgi protein 1; sorting nexin-18; Sorting nexin-associated Golgi protein 1
Gene Symbols SNX18
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N terminal region of human SNAG1 (aa 107-156).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112574
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.