missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNX18 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142049
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members, but contains a SH3 domain. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| SNX18 | |
| Polyclonal | |
| Unconjugated | |
| SNX18 | |
| SH3 and PX domain-containing protein 3B; SH3PX2; SH3PXD3B; Snag1; Snx18; sorting nexin 18; sorting nexin associated golgi protein 1; sorting nexin-18; Sorting nexin-associated Golgi protein 1 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 112574 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q96RF0 | |
| SNX18 | |
| Peptide with sequence C-KVQRVPLMTVLSF. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction