missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX2 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21430AF532
This item is not returnable.
View return policy
Description
SNX2 Polyclonal antibody specifically detects SNX2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| SNX2 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| MGC5204, sorting nexin 2, sorting nexin-2, Transformation-related gene 9 protein, TRG-9 | |
| Produced in rabbits immunized with E. coli-derived Human SNX2 fragment | |
| 0.1 mL | |
| Signal Transduction | |
| 6643 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction