missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX20 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13180-1-AP-150UL
This item is not returnable.
View return policy
Description
SNX20 interacts with the cytoplasmic domain of PSGL1 and cycles PSGL1 into endosomes.Specifications
| SNX20 | |
| Polyclonal | |
| Unconjugated | |
| SNX20 | |
| SLIC 1, SLIC1, SNX20, sorting nexin 20 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 124460, 307742, 71607 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5BK61, Q7Z614, Q9D2Y5 | |
| Snx20 | |
| SNX20 Fusion Protein Ag3847 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction