missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SNX24 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15999974
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15999974 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15999974 Supplier Invitrogen™ Supplier No. PA567315

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MEVYIPSFRYE ESDLERGYTV FKIEVLMNGR KHFVEKRYSE FHA Highest antigen sequence identity to the following orthologs - mouse 95%, rat 95%.

May be involved in several stages of intracellular trafficking.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX24
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SNX24
Gene Accession No. Q9Y343
Gene Alias 2810011K15Rik; 5730433I16Rik; AI451341; PRO1284; SBBI31; Snx24; sorting nexin 24; sorting nexin-24; sorting nexing 24; UNQ654/PRO1284
Gene Symbols SNX24
Host Species Rabbit
Immunogen Recombinant Human SNX24. Recombinant protein control fragment (Product #RP-107956).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28966
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.