missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SNX27 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29174
This item is not returnable.
View return policy
Description
SNX27 Polyclonal antibody specifically detects SNX27 in Human,Mouse samples. It is validated for Western Blot
Specifications
| SNX27 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| KIAA0488MGC126873, methamphetamine-responsive transcript 1, MGC126871, MGC20471, MRT1, MY014, sorting nexin family member 27, sorting nexin-27 | |
| Between 125 and 175 | |
| 100 μg | |
| Signal Transduction | |
| 81609 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction