missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNX30 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570806
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
SNX9, SNX18 and SNX30 constitute a separate subfamily of PX-BAR-containing sorting nexin (SNX) proteins. May be involved in several stages of intracellular trafficking.
Specifications
| SNX30 | |
| Polyclonal | |
| Unconjugated | |
| SNX30 | |
| 4732481H14Rik; ATG24A; C030041J06Rik; hypothetical protein LOC550496; novel PX domain containing protein; RGD1311367; Snx30; sorting nexin family member 30; sorting nexin-30; zgc:112424 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 209131 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8CE50 | |
| SNX30 | |
| synthetic peptide directed towards the middle region of mouse SNX30. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction