missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNX33 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559378
This item is not returnable.
View return policy
Description
Immunogen sequence: VRSGVEAFIL GDVPMMAKIA ETYSIEMGPR GPQWKANPHP FACSVEDPTK QTK Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
SNX33 (sorting nexin-33), also known as SH3PX3, SH3PXD3C or SNX30, is a 574 amino acid protein that interacts with ADAM15 and FAS-L. Belonging to the sorting nexin family, SNX33 contains one BAR domain, one PX (phox homology) domain and one SH3 domain. The gene that encodes SNX33 consists of over 14,000 bases and maps to human chromosome 15q24.2. Housing approximately 106 million base pairs and encoding more than 700 genes, chromosome 15 makes up about 3% of the human genome. Angelman and Prader-Willi syndromes are associated with loss of function or deletion of genes in the 15q11-q13 region.
Specifications
| SNX33 | |
| Polyclonal | |
| Unconjugated | |
| SNX33 | |
| SH3 and PX domain containing 3; SH3 and PX domain-containing protein 3; SH3PX3; SH3PXD3C; SNX30; SNX33; sorting nexin 33; sorting nexin-33 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 257364 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WV41 | |
| SNX33 | |
| Recombinant protein corresponding to Human SNX33. Recombinant protein control fragment (Product #RP-98138). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction