missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX5 Antibody (SR1821), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22592-50ul
This item is not returnable.
View return policy
Description
SNX5 Monoclonal antibody specifically detects SNX5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications
| SNX5 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| SR1821 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| FLJ10931, sorting nexin 5, sorting nexin-5 | |
| A synthesized peptide derived from human SNX5 (Uniprot #: Q9Y5X3) | |
| 50 μg | |
| Core ESC Like Genes, Signal Transduction, Stem Cell Markers | |
| 27131 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction