missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Snx6 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94535-0.1ml
This item is not returnable.
View return policy
Description
Snx6 Polyclonal antibody specifically detects Snx6 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Snx6 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| MGC3157, MSTP010, sorting nexin 6, sorting nexin-6, TFAF2, TRAF4-associated factor 2, tumor necrosis factor receptor-associated factor 4(TRAF4)-associated factor 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-290 of human SNX6 (NP_067072.3). MTKEEFTKMKQELEAEYLAIFKKTVAMHEVFLCRVAAHPILRRDLNFHVFLEYNQDLSVRGKNKKEKLEDFFKNMVKSADGVIVSGVKDVDDFFEHERTFLLEYHNRVKDASAKSDRMTRSHKSAADDYNRIGSSLYALGTQDSTDICKFFLKVSELFDKTRKIEARVSADEDLKLSDLLKYYLRESQAAKDLLYRRSRSLVDYENANKALDKARAKNKDVLQAETSQQLCCQKFEKISESAKQELIDFKTRRVAAFRKNLVELAELELKHAKGNLQLLQNCLAVLNGDT | |
| 0.1 mL | |
| Signal Transduction | |
| 58533 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction