missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX6, Mouse anti-Human, Clone: 3B11, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00058533-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MTKEEFTKMKQELEAEYLAIFKKTVAMHEVFLCRVAAHPILGRDLNFHVFLEYNQDLSVRGKNKKEKLEDFFKNMVKSADGVIVSGVKDVDDFFEHERTFLLEYHNRVKDASAKSDRMTRSHKSAADDYNRIGSSLYALGTQDSTDICKFFLKVSELFDKTRKIEARVSADEDLKLSDLLKYYLRESQAAKDLLYRRSRSLVDYENANKALDKARAKNKDVLQAETSQQLCCQKFEKISESAKQELIDFKTRRVAAFRKNLVELAELELKHAKGNLQLLQNCLAVLNGDTSpecifications
| SNX6 | |
| Monoclonal | |
| Unconjugated | |
| sorting nexin 6 | |
| MGC3157/MSTP010/TFAF2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 3B11 | |
| In 1x PBS, pH 7.4 | |
| BC001798 | |
| SNX6 | |
| SNX6 (AAH01798, 1 a.a. ∼ 290 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 58533 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction