missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX8 Antibody (OTI1A1), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74265UV
This item is not returnable.
View return policy
Description
SNX8 Monoclonal specifically detects SNX8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| SNX8 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| SNX8 | |
| Full length recombinant protein of human SNX8 (NP_037453) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1A1 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| DKFZp761E1721, Mvp1, sorting nexin 8, sorting nexin-8 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 29886.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction