missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SNX8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5118275
This item is not returnable.
View return policy
Description
SNX8 Polyclonal Antibody for Western Blot, IHC (P), IP
Sorting nexin (SNX) proteins are members of a large family of hydrophilic PX (phospholipid-binding motif) domain-containing proteins that interact with a variety of receptor types. SNXs are widely expressed, although the tissue distribution of each SNX mRNA varies. The ability of SNXs to bind specific phospholipids, as well as their tendency to form protein-protein complexes, suggests a role for these proteins in cellular membrane trafficking and protein sorting. SNXs may also function specifically in pro-degradative sorting, internalization, endosomal recycling or simply in endosomal sorting. SNXs partially associate with cellular membranes, despite their hydrophilic nature. SNX8 is an ortholog of a yeast protein.
Specifications
| SNX8 | |
| Polyclonal | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q9Y5X2 | |
| Snx8 | |
| E. coli-derived Human SNX8 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Snx8 | |
| B130023O14Rik; Mvp1; SNX8; sorting nexin 8; sorting nexin-8 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 29886 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction