missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SNX9 Polyclonal Antibody

Product Code. 13492007
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13492007 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13492007 Supplier Invitrogen™ Supplier No. PA527896

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: H1299, HCT116. Predicted reactivity: Mouse (95%), Rat (94%), Xenopus laevis (84%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members, but does contain a SH3 domain near its N-terminus. This protein interacts with the cytoplasmic domains of the precursor but not the processed forms of a disintegrin and metalloprotease domain 9 and 15. This protein binds the beta-appendage domain of adaptor protein 2 and may function to assist adaptor protein 2 in its role at the plasma membrane. This protein interacts with activated Cdc42-associated kinase-2 to regulate the degradation of epidermal growth factor receptor protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SNX9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Snx9
Gene Accession No. Q9Y5X1
Gene Alias 2700073N08Rik; MST155; MSTP155; OTTHUMP00000040083; Protein SDP1; RP11-266C7.10; SDP1; SH3 and PX domain-containing protein 1; SH3 and PX domain-containing protein 3A; SH3PX1; SH3PXD3A; Snx9; sorting nexin 9; sorting nexin-9; Wiskott-Aldrich syndrome protein (WASP) interactor protein; WISP
Gene Symbols Snx9
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SNX9. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51429
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.