Learn More
SOAT2 Mouse anti-Human, Clone: OTI2D3, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF800775
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene is a member of a small family of acyl coenzyme A:cholesterol acyltransferases. The gene encodes a membrane-bound enzyme localized in the endoplasmic reticulum that produces intracellular cholesterol esters from long-chain fatty acyl CoA and cholesterol. The cholesterol esters are then stored as cytoplasmic lipid droplets inside the cell. The enzyme is implicated in cholesterol absorption in the intestine and in the assembly and secretion of apolipoprotein B-containing lipoproteins such as very low density lipoprotein (VLDL). Several alternatively spliced transcript variants of this gene have been described, but their full-length nature is not known.Specifications
| SOAT2 | |
| Monoclonal | |
| Unconjugated | |
| SOAT2 | |
| ACACT2, ACAT2, ARGP2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 8435 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin) | |
| OTI2D3 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O75908 | |
| SOAT2 | |
| Human recombit protein fragment corresponding to amino acids 1-120 of human SOAT2 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.