Learn More
SOCS-3 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-27210AF532
Product Code. 18107892
  • € 577.00 / 0.10mL
Description
SOCS-3 Polyclonal specifically detects SOCS-3 in Human, Feline, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SOCS-3
Polyclonal
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
CIS3CIS-3, Cish3, cytokine-induced SH2 protein 3, Cytokine-inducible SH2 protein 3, SOCS-3STAT-induced STAT inhibitor 3, SSI-3ATOD4, SSI3MGC71791, suppressor of cytokine signaling 3
Rabbit
Affinity Purified
RUO
Primary
Store at 4°C in the dark.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 532
50mM Sodium Borate with 0.05% Sodium Azide
SOCS3
A portion of amino acids 110-160 from human SOCS3 was used as the immunogen for this antibody.
0.1 mL
Signal Transduction
Human, Feline, Primate