missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOCS-3 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27210B
This item is not returnable.
View return policy
Description
SOCS-3 Polyclonal specifically detects SOCS-3 in Human, Feline, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SOCS-3 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CIS3CIS-3, Cish3, cytokine-induced SH2 protein 3, Cytokine-inducible SH2 protein 3, SOCS-3STAT-induced STAT inhibitor 3, SSI-3ATOD4, SSI3MGC71791, suppressor of cytokine signaling 3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| SOCS3 | |
| A portion of amino acids 110-160 from human SOCS3 was used as the immunogen for this antibody. | |
| 0.1 mL | |
| Signal Transduction | |
| Human, Feline, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction