missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse IL-19 Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
Description
IL-19 protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using human IL-20Ra and human IL-20Rb co-transfected murine BaF3 pro-B cells is less than 0.8 ng/mL, corresponding to a specific activity of > 1.25 x 10^6 IU/mg.
Specifications
Specifications
| For Use With (Application) | Western Blot, Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4, 3% Trehalose. |
| Gene ID (Entrez) | 29949 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 17.6 kDa |
| Quantity | 10 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin Concentration | Less than 0.1 EU/μg of IL-19 AS determined by LAL method. |
| Gene Alias | IL-10 C, IL-19, interleukin 19, MDA1, melanoma differentiation associated protein-like protein, NG0.1 Melanoma differentiation-associated protein-like protein, ZMDA1interleukin-19 |
| Gene Symbol | IL19 |
| Reconstitution | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction