missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOCS-4 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-898
This item is not returnable.
View return policy
Description
SOCS-4 Polyclonal specifically detects SOCS-4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| SOCS-4 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| CIS4, DKFZp686J1568, SOCS-4, SOCS7, SOCS-7, SSI4, STAT4, suppressor of cytokine signaling 4, Suppressor of cytokine signaling 7SH2 domain containing SOCS box protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Both variants represent identical protein (NP_543143.1 and NP_955453.1). | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Immunohistochemistry 3 - 6 ug/ml, Immunohistochemistry-Paraffin 3 - 6 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| NP_543143. | |
| SOCS4 | |
| Peptide with sequence C-SKVRVLRIDAPEQQ corresponding to C-Terminus according to NP_543143.1, NP_955453.1. | |
| 0.1 mg | |
| Cell Biology, Signal Transduction | |
| 122809 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction