missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SOCS4 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5141999
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
The protein encoded by this gene contains a SH2 domain and a SOCS BOX domain. The protein thus belongs to the suppressor of cytokine signaling (SOCS), also known as STAT-induced STAT inhibitor (SSI), protein family. SOCS family members are known to be cytokine-inducible negative regulators of cytokine signaling. Two alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
| SOCS4 | |
| Polyclonal | |
| Unconjugated | |
| SOCS4 | |
| 3110032M18Rik; A730004F22Rik; AI427843; AU015727; AU015859; SH2 domain containing SOCS box protein; SOCS4; SOCS-4; SOCS7; SOCS-7; suppressor of cytokine signaling 4; Suppressor of cytokine signaling 7; suppressor of cytokine signalling 4 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 122809 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q8WXH5 | |
| SOCS4 | |
| Peptide with sequence C-SKVRVLRIDAPEQQ (aa 426-439). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction