missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SOCS5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA141253
This item is not returnable.
View return policy
Description
Suggested positive control: human spleen protein, human or mouse spleen, mouse spleen protein.
Accumulating evidence demonstrates that cytokine receptor signaling is negatively regulated by a family of Src homology 2 domain-containing adaptor molecules Termed SOCS (Suppressor of Cytokine Signaling). To date, there are eight members of SOCS family have been recognized, they are SOCS-1, 2, 3, 4, 5, 6, 7 and CIS. Structurally the SOCS proteins are composed of an N-terminal region of variable length and amino acid composition, a central SH2 domain, and a previously unrecognized C-terminal motif that we have called the SOCS box. The SOCS proteins appear to form part of a classical negative feed back loop that regulates cytokine signal transduction via a STAT-induced transcriptional mechanism. Transcription of each of the SOCS genes occurs rapidly in vitro and in vivo in response to cytokines, and once produced, the various members of the SOCS family appear to inhibit signaling in different ways.
Specifications
| SOCS5 | |
| Polyclonal | |
| Unconjugated | |
| SOCS5 | |
| 1810018L08Rik; CIS6; CIS-6; CISH5; CISH6; cytokine inducible SH2-containing protein 5; cytokine-inducible SH2 protein 6; Cytokine-inducible SH2-containing protein 5; KIAA0671; mKIAA0671; RGD1564914; Socs5; SOCS-5; Suppressor of cytokine signaling 5 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 500616, 56468, 9655 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.10 mg/mL | |
| PBS with 100μg/mL BSA and 0.02% sodium azide | |
| O54928, O75159 | |
| SOCS5 | |
| Synthetic peptide containing amino acid residues of human SOCS5 protein. | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction