missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOD2/Mn-SOD Antibody, CoraFluor™ 1, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB200-165CL1
This item is not returnable.
View return policy
Description
SOD2/Mn-SOD Polyclonal antibody specifically detects SOD2/Mn-SOD in Rat samples. It is validated for Western Blot
Specifications
| SOD2/Mn-SOD | |
| Polyclonal | |
| PBS | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
| Western Blot | |
| CoraFluor 1 | |
| EC 1.15.1.1, indophenoloxidase B, IPOB, manganese-containing superoxide dismutase, mangano-superoxide dismutase, Mn superoxide dismutase, MNSOD, MVCD6, superoxide dismutase [Mn], mitochondrial, superoxide dismutase 2, mitochondrial | |
| A synthetic peptide between amino acids 175-222 of human manganese superoxide dismutase encoded within exon 5 (LocusLink ID 6648). | |
| 0.1 mL | |
| Cancer, Diabetes Research, Neuroscience, Tumor Suppressors | |
| 6648 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction