missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOD2/Mn-SOD Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
€ 297.00
Specifications
| Antigen | SOD2/Mn-SOD |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:5000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
SOD2/Mn-SOD Polyclonal specifically detects SOD2/Mn-SOD in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Guinea Pig, Hamster, Gerbil, Monkey, Rabbit, Sheep, Xenopus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
| SOD2/Mn-SOD | |
| Western Blot 1:5000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Diabetes Research, Tumor Suppressors | |
| PBS (pH 7.4), 50% Glycerol with 0.09% Sodium Azide | |
| EC 1.15.1.1, indophenoloxidase B, IPOB, manganese-containing superoxide dismutase, mangano-superoxide dismutase, Mn superoxide dismutase, MNSOD, MVCD6, superoxide dismutase [Mn], mitochondrial, superoxide dismutase 2, mitochondrial | |
| SOD2 | |
| IgG | |
| Affinity Purified | |
| Detects approx 25 kDa. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| P04179 | |
| 6648 | |
| Recombinant Human Mn SOD Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title