missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOD3/EC-SOD Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18234AF647
This item is not returnable.
View return policy
Description
SOD3/EC-SOD Polyclonal antibody specifically detects SOD3/EC-SOD in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| SOD3/EC-SOD | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| EC-SODEC 1.15.1.1, extracellular superoxide dismutase [Cu-Zn], MGC20077, superoxide dismutase 3, extracellular | |
| Peptide corresponding to AA 227-236 of human EC SOD | |
| 0.1 mL | |
| Cancer, Neurodegeneration, Neuroscience | |
| 6649 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction