missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOD3/EC-SOD Antibody (SR1423), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21798-100ul
This item is not returnable.
View return policy
Description
SOD3/EC-SOD Monoclonal antibody specifically detects SOD3/EC-SOD in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| SOD3/EC-SOD | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1423 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| EC-SODEC 1.15.1.1, extracellular superoxide dismutase [Cu-Zn], MGC20077, superoxide dismutase 3, extracellular | |
| A synthesized peptide derived from human SOD3/EC-SOD (Uniprot #: P08294) | |
| 100 μg | |
| Cancer, Neurodegeneration, Neuroscience | |
| 6649 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction