Learn More
Invitrogen™ SOD3 Monoclonal Antibody (4GG11G6), APC
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA545223
Product Code. 17838692
  • € 455.00 / 100µg
Description
1 μg/mL of MA5-45223 was sufficient for detection of EC-SOD in 20 μg of human cartilage lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects extracellular SOD approximately 35kDa.
This gene encodes a member of the vaccinia-related kinase (VRK) family of serine/threonine protein kinases. This gene is widely expressed in human tissues and has increased expression in actively dividing cells, such as those in testis, thymus, fetal liver, and carcinomas. Its protein localizes to the nucleus and has been shown to promote the stability and nuclear accumulation of a transcriptionally active p53 molecule and, in vitro, to phosphorylate Thr18 of p53 and reduce p53 ubiquitination. This gene, therefore, may regulate cell proliferation. This protein also phosphorylates histone, casein, and the transcription factors ATF2 (activating transcription factor 2) and c-JUN.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SOD3
Monoclonal
1 mg/mL
2.48mM MES, 95.46mM phosphate with 2mM EDTA and no preservative; pH 7.4
O09164, P08294, Q08420
SOD3
Human extracellular SOD purified from aortas.
100 μg
Primary
Human, Mouse, Rat, Guinea Pig
Antibody
IgG1 κ
ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry
4GG11G6
APC
SOD3
AI314465; ALS; ALS1; EC SOD; EC-SOD; ECSODPT; extracellular superoxide dismutase; extracellular superoxide dismutase [Cu-Zn]; MGC20077; SOD 3; SOD3; Sod-3; Superoxide dimutase 3; superoxide dismutase; superoxide dismutase 3; superoxide dismutase 3, extracellular; superoxide dismutase B; testicular tissue protein Li 175
Mouse
Protein G
RUO
100718762, 20657, 25352, 6649
4°C