missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SOD3 Monoclonal Antibody (4GG11G6), PE

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545226
This item is not returnable.
View return policy
Description
1 μg/mL of MA5-45226 was sufficient for detection of EC-SOD in 20 μg of human cartilage lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects extracellular SOD approximately 35kDa.
This gene encodes a member of the vaccinia-related kinase (VRK) family of serine/threonine protein kinases. This gene is widely expressed in human tissues and has increased expression in actively dividing cells, such as those in testis, thymus, fetal liver, and carcinomas. Its protein localizes to the nucleus and has been shown to promote the stability and nuclear accumulation of a transcriptionally active p53 molecule and, in vitro, to phosphorylate Thr18 of p53 and reduce p53 ubiquitination. This gene, therefore, may regulate cell proliferation. This protein also phosphorylates histone, casein, and the transcription factors ATF2 (activating transcription factor 2) and c-JUN.
Specifications
| SOD3 | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
| O09164, P08294, Q08420 | |
| SOD3 | |
| Human extracellular SOD purified from aortas. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Guinea Pig | |
| Antibody | |
| IgG1 κ |
| ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 4GG11G6 | |
| PE | |
| SOD3 | |
| AI314465; ALS; ALS1; EC SOD; EC-SOD; ECSODPT; extracellular superoxide dismutase; extracellular superoxide dismutase [Cu-Zn]; MGC20077; SOD 3; SOD3; Sod-3; Superoxide dimutase 3; superoxide dismutase; superoxide dismutase 3; superoxide dismutase 3, extracellular; superoxide dismutase B; testicular tissue protein Li 175 | |
| Mouse | |
| Protein G | |
| RUO | |
| 100718762, 20657, 25352, 6649 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction