missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SOD3 Monoclonal Antibody (C1)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA543570

 View more versions of this product

Product Code. 17664026

  • € 180.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SOD3 Monoclonal Antibody for Western Blot

This gene encodes a member of the vaccinia-related kinase (VRK) family of serine/threonine protein kinases. This gene is widely expressed in human tissues and has increased expression in actively dividing cells, such as those in testis, thymus, fetal liver, and carcinomas. Its protein localizes to the nucleus and has been shown to promote the stability and nuclear accumulation of a transcriptionally active p53 molecule and, in vitro, to phosphorylate Thr18 of p53 and reduce p53 ubiquitination. This gene, therefore, may regulate cell proliferation. This protein also phosphorylates histone, casein, and the transcription factors ATF2 (activating transcription factor 2) and c-JUN.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SOD3
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
P08294
SOD3
Recombinant protein Superoxide Dismutase 3, Extracellular. The antigen corresponds to amino acid range 19-240 of the target protein.
20 μL
Primary
Human
Antibody
IgG1 κ
Western Blot
C1
Unconjugated
SOD3
AI314465; ALS; ALS1; EC SOD; EC-SOD; ECSODPT; extracellular superoxide dismutase; extracellular superoxide dismutase [Cu-Zn]; MGC20077; SOD 3; SOD3; Sod-3; Superoxide dimutase 3; superoxide dismutase; superoxide dismutase 3; superoxide dismutase 3, extracellular; superoxide dismutase B; testicular tissue protein Li 175
Mouse
Protein A/G
RUO
6649
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.