Learn More
Witeg™ Soda Glass Evaporating Dish
Evaporate solvents in a sample using the Witeg™ Soda Glass Evaporating Dish, typically used to accommodate small samples.
Brand: Witeg™ 5 600 080
Additional Details : Weight : 0.02300kg
Description
- Watch glass shape
- Thin glass
- Fire-polished rim
- Soda glass
Specifications
Soda Lime Glass | |
Evaporating Dish | |
80 mm |