Learn More
Witeg™ Soda Glass Evaporating Dish
Evaporate solvents in a sample using the Witeg™ Soda Glass Evaporating Dish, typically used to accommodate small samples.
Brand:  Witeg™ 5 600 080
Additional Details : Weight : 0.02300kg
 View more versions of this product
Product Code. 10787545
  • € 122.00 / Pack of 10
Estimated Shipment: 21-05-2024
to see stock.
Add to basket
Description
Description
  • Watch glass shape
  • Thin glass
  • Fire-polished rim
  • Soda glass
Specifications
Specifications
Soda Lime Glass
Evaporating Dish
80 mm