Learn More
Gravis Glass Sterilizing Jars
Catalog Number p-8008467
Click to view available options
Capacity (Metric):
500 mL
750 mL
1000 mL
2000 mL
Height (Metric):
108 mm
145 mm
178 mm
248 mm
Providing secure storage and closure for sterilizing purposes in the laboratory. Gravis Glass Sterilizing Jars are provided with a metal locking bracket and rubber seal around the rim.
  • Manufactured from soda lime glass
  • Available in a range of capacities
Closure Type Clip
Diameter (Metric) 98 mm
Height (Metric) 108 mm
Product Type Jar
Capacity (Metric) 500 mL
Diameter (Metric) Outer Neck 90 mm
Includes Metal clip closure and rubber seal
Material Soda Lime Glass
Quantity 6Pack