missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Soda Lime Glass Light Wall Tubing

Manufactured from soda lime glass, this soda lime glass light walled tubing are ideal in laboratory, scientific research, and other environments.

Brand:  Fisherbrand™

Additional Details : Weight : 0.50000kg

Product Code. 10653615

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Tolerance ±0.2mm; wall thickness 0.7mm
  • Manufactured from soda lime glass
Specifications

Specifications

Soda Lime Glass
4 mm
50 Pack
±0.2mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers