Learn More
Lenz Laborglasinstrumente™ Plain Funnels
Product Code. 10366021
Click to view available options
Length (Metric) Stem:
30mm
35mm
40mm
45mm
50mm
55mm
60mm
70mm
80mm
100mm
Unit Size:
Pack of 10
Product Code. 10366021
Brand: Lenz Laborglasinstrumente™ 05318020
Only null left
Add to basket
Add to basket
For the safe and easy mixing, pouring and storage of chemicals in the laboratory. Lenz Laborglasinstrumente™ Plain Funnels are manufactured from soda-lime glass.
  • With short stem
  • Angle at 60°
Specifications
Diameter (Metric) Stem 9mm
Material Soda Lime Glass
Angle 60°
Length (Metric) Stem 70mm