Learn More
Hirschmann™ Soda-lime Glass Test Tube with Round Bottoms
Product Code. 10528771
Click to view available options
Diameter (Metric):
12 mm
16 mm
Length (Metric):
75 mm
100 mm
125 mm
150 mm
160 mm
180 mm
Unit Size:
Pack of 100
Product Code. 10528771
Brand: Hirschmann™ 9270106
Only null left
Add to basket
Add to basket
Test tubes without scale made from sodalime glass.
  • Plain end
  • Round bottom
  • Sodalime-glass
Specifications
Description Dimensions: 75 x 12 mm, Wall thickness: 0.8 mm
Length (Metric) 75 mm
Material Soda Lime Glass
Quantity 100/Pk.
Diameter (Metric) 12 mm
Product Type Test Tube