Learn More
Scientific Glass Labs™ Soda Lime Glass Watch Glasses
Product Code. 13142440
Change view
Click to view available options
Diameter (Metric):
70 mm
80 mm
150 mm
Unit Size:
Pack of 10
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
13142440 150 mm Pack of 5
13102440 80 mm Pack of 10
13192430 70 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 13142440 Supplier Scientific Glass Labs™ Supplier No. 170/1/150
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
Manufactured from Soda lime glass. Scientific Glass Labs™ Soda Lime Glass Watch Glasses with thin, clear glass and ground edge can be used to evaporate liquids, hold solids for weighing, or for heating small substances.
Watch glasses are useful for evaporating liquids, holding solids for weighing, or for heating small substances.
  • Manufactured from Soda lime glass
  • Thin, clear glass
  • Ground edge
Specifications
Bottom Shape Flat
Material Soda Lime Glass
Shape Round
Type Watch Glass
For Use With (Application) Evaporating Liquids, Heating Small Substances, Holding Solids for Weighing
Interior Finish Ground Edge
Diameter (Metric) 150 mm