Learn More
Sodium 2-Keto-4(RS)-methyl-d3-pentanoate, CDN
Specifications
Specifications
| Chemical Name or Material | 4-Methyl-2-oxopentanoic acid sodium salt methyl-d3 |
| CAS | 93523-67-2 |
| Molecular Formula | C6 2H3 H6 O3 . Na |
| Quantity | 0.5 g |
| Synonym | Pentanoic-5,5,5-d3 acid, 4-methyl-2-oxo-, sodium salt (9 CI), 4-Methyl-2-oxopentanoic-5,5,5-D3 acid sodium salt, 2-Oxo-4-(methyl-d3)pentanoic acid sodium salt, 2-Oxo-4-methylpentanoic acid-d3 sodium salt, 4-(Methyl-d3)-2-oxopentanoic acid sodium salt, Sodium 2-ketoisocaproate-d3, Sodium 4-methyl-2-oxopentanoate-5,5,5-d3, Sodium 4-methyl-2-oxovalerate-d3, Sodium α-keto-isocaproate-d3, Sodium α-oxoisocaproate-delta;,delta;,delta;-d3 |
| InChI Formula | InChI=1 S/C6H10O3.Na/c1-4(2)3-5(7)6(8)9;/h4H,3H2,1-2H3,(H,8,9);/q;+1/p-1/i1D3; |
| SMILES | [Na+].[2 H]C([2 H])([2 H])C(C)CC(=O)C(=O)[O-] |
| IUPAC Name | sodium;5,5,5-trideuterio-4-methyl-2-oxopentanoate |
| Molecular Weight (g/mol) | 155.14 |
| Formula Weight | 155.0638 g/mol |
| Show More |