missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-Decylbenzenesulfonate, TRC

Product Code. 30413682
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30413682 100 mg 100mg
30414781 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30413682 Supplier Toronto Research Chemicals Supplier No. TRCS622000100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2627-06-7
Molecular Formula C16 H25 O3 S . Na
Molecular Weight (g/mol) 320.42
Synonym Benzenesulfonic acid, 4-decyl-, sodium salt (9CI), Benzenesulfonic acid, p-decyl-, sodium salt (6CI,7CI,8CI), Sodium 4-decylbenzenesulfonate, Sodium p-decylbenzenesulfonate, p-Decylbenzenesulfonic acid sodium salt
IUPAC Name sodium;4-decylbenzenesulfonate
SMILES [Na+].CCCCCCCCCCc1ccc(cc1)S(=O)(=O)[O-]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C16H26O3S.Na/c1-2-3-4-5-6-7-8-9-10-15-11-13-16(14-12-15)20(17,18)19;/h11-14H,2-10H2,1H3,(H,17,18,19);/q;+1/p-1
Formula Weight 320.1422
Chemical Name or Material 4-n-Decylbenzenesulfonic Acid Sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.